siRNA Details
| Gene Symbol: TTC38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017931.2 | NP_060401.2 | 2, 3 | 191 | ||
| XM_011530260.2 | XP_011528562.1 | 2, 3 | 191 | |||
| GenBank mRNA | AK000706.1 | - | 2, 3 | 147 | ||
| AK300606.1 | - | 2, 3 | 171 | |||
| BC018918.2 | - | 2, 3 | 132 | 4130906 | ||
| DA762669.1 | - | 2, 3 | 193 | |||

TTC38
Genomic Map