siRNA Details
| Gene Symbol: PID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100818.1 | NP_001094288.1 | 2, 3 | 483 | ||
| NM_017933.4 | NP_060403.3 | 3, 4 | 694 | |||
| XM_011511384.2 | XP_011509686.1 | 2, 3 | 366 | |||
| XM_017004404.1 | XP_016859893.1 | 3, 4 | 5625 | |||
| XM_017004405.1 | XP_016859894.1 | 4, 5 | 564 | |||
| XM_017004406.1 | XP_016859895.1 | 2, 3 | 222 | |||
| GenBank mRNA | AB075874.1 | - | 3, 4 | 349 | ||
| AK000708.1 | - | 469 | ||||
| AK096636.1 | - | 2, 3 | 425 | |||
| AK125359.1 | - | 3, 4 | 618 | |||
| AY317148.1 | - | 2, 3 | 315 | |||
| DB474424.1 | - | 2, 3 | 483 | |||

PID1
Genomic Map