siRNA Details
| Gene Symbol: PTCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017952.5 | NP_060422.4 | 8 | 680 | ||
| GenBank mRNA | AK293962.1 | - | 7 | 499 | ||
| AK315676.1 | - | 2 | 134 | |||
| BC011832.2 | - | 8 | 615 | 3939273 | ||
| BI825315.1 | - | 8 | 668 | |||

PTCD3
Genomic Map