siRNA Details
| Gene Symbol: PTCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017952.5 | NP_060422.4 | 24 | 2293 | ||
| GenBank mRNA | AK000765.1 | - | 14 | 1463 | ||
| AL832520.1 | - | 13 | 1296 | |||
| BC001758.1 | - | 13 | 1366 | |||
| BC011832.2 | - | 24 | 2227 | 3939273 | ||

PTCD3
Genomic Map