siRNA Details
| Gene Symbol: SRBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018079.4 | NP_060549.4 | 3, 4 | 342 | ||
| XM_011532946.1 | XP_011531248.1 | 3, 4 | 343 | |||
| XM_017004417.1 | XP_016859906.1 | 3, 4 | 343 | |||
| GenBank mRNA | AK056536.1 | - | 3, 4 | 312 | ||
| DA235970.1 | - | 3, 4 | 343 | |||

SRBD1
Genomic Map