siRNA Details
| Gene Symbol: SRBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018079.4 | NP_060549.4 | 21 | 3128 | ||
| XM_011532946.1 | XP_011531248.1 | 21 | 3081 | |||
| XM_017004418.1 | XP_016859907.1 | 13 | 2682 | |||
| GenBank mRNA | AK001241.1 | - | 15 | 2025 | ||
| AK055896.1 | - | 6 | 1948 | |||
| AK056536.1 | - | 21 | 3098 | |||
| AK301122.1 | - | 13 | 1827 | |||
| AK316222.1 | - | 13 | 1808 | |||
| BC032538.1 | - | 15 | 2103 | 5528417 | ||

SRBD1
Genomic Map