siRNA Details
| Gene Symbol: ANKZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042410.1 | NP_001035869.1 | 8 | 979 | ||
| NM_001282792.1 | NP_001269721.1 | 5 | 658 | |||
| NM_018089.2 | NP_060559.2 | 8 | 1052 | |||
| XM_005246663.2 | XP_005246720.1 | 8 | 971 | |||
| XM_011511392.2 | XP_011509694.1 | 8 | 1052 | |||
| XM_017004420.1 | XP_016859909.1 | 8 | 971 | |||
| XM_017004421.1 | XP_016859910.1 | 8 | 1052 | |||
| GenBank mRNA | AF364318.1 | - | 8 | 906 | ||
| AK001277.1 | - | 8 | 989 | |||
| AK023206.1 | - | 8 | 979 | |||
| AK095304.1 | - | 8 | 916 | |||
| AK098282.1 | - | 6 | 2033 | |||
| AK225275.1 | - | 8 | 977 | |||
| AK225309.1 | - | 8 | 953 | |||
| AK303077.1 | - | 7 | 785 | |||
| AK303537.1 | - | 5 | 577 | |||
| BC000238.1 | - | 8 | 934 | 3351742 | ||
| BC008948.2 | - | 8 | 963 | 2959748 | ||
| BC028334.1 | - | 5 | 580 | |||
| DA899467.1 | - | 2 | 215 | |||
| DQ412564.1 | - | 8 | 989 | |||

ANKZF1
Genomic Map