siRNA Details
| Gene Symbol: TMEM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018126.2 | NP_060596.2 | 7 | 1451 | ||
| XM_005248116.3 | XP_005248173.1 | 8 | 1290 | |||
| XM_005248117.2 | XP_005248174.1 | 8 | 1238 | |||
| GenBank mRNA | AK001387.1 | - | 7 | 1419 | ||
| AK023062.1 | - | 7 | 1431 | |||
| AK024587.1 | - | 1 | 159 | |||
| AK094190.1 | - | 8 | 1364 | |||
| AK308501.1 | - | 6 | 1231 | |||
| AY659966.1 | - | 7 | 1419 | |||
| BC000948.2 | - | 7 | 1181 | 3448888 | ||

TMEM33
Genomic Map