siRNA Details
| Gene Symbol: PNMAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103149.1 | NP_001096619.1 | 2 | 1074 | ||
| NM_018215.3 | NP_060685.2 | 2 | 1074 | |||
| XM_011527067.2 | XP_011525369.1 | 2 | 1088 | |||
| GenBank mRNA | AK001643.1 | - | 1 | 93 | ||
| AK096326.1 | - | 2 | 634 | |||
| AK290218.1 | - | 3 | 957 | |||
| AL834344.1 | - | 2 | 918 | |||
| BC026026.1 | - | 1 | 107 | |||
| BC032508.1 | - | 2 | 1052 | 5246377 | ||
| BC051688.1 | - | 2 | 1031 | 5295669 | ||
| BX649091.1 | - | 1 | 169 | |||

PNMAL1
Genomic Map