siRNA Details
| Gene Symbol: PNMAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103149.1 | NP_001096619.1 | 3 | 3435 | ||
| NM_018215.3 | NP_060685.2 | 3 | 3640 | |||
| XM_011527067.2 | XP_011525369.1 | 3 | 3725 | |||
| GenBank mRNA | AK001643.1 | - | 2 | 2665 | ||
| AK096326.1 | - | 3 | 3204 | |||
| AL834344.1 | - | 3 | 3279 | |||
| BC026026.1 | - | 2 | 2677 | |||
| BC032508.1 | - | 3 | 3624 | 5246377 | ||
| BC051688.1 | - | 3 | 3603 | 5295669 | ||
| BX649091.1 | - | 2 | 2478 | |||

PNMAL1
Genomic Map