siRNA Details
| Gene Symbol: TMEM39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018266.2 | NP_060736.1 | 9 | 1713 | ||
| NR_073506.1 | 10 | 1822 | ||||
| XM_005247578.1 | XP_005247635.1 | 8 | 1557 | |||
| XM_006713687.1 | XP_006713750.1 | 9 | 1621 | |||
| XM_017006788.1 | XP_016862277.1 | 8 | 1594 | |||
| GenBank mRNA | AK001764.1 | - | 9 | 1655 | ||
| AK222950.1 | - | 9 | 1645 | |||
| AK223248.1 | - | 9 | 1645 | |||
| BC003192.1 | - | 10 | 1737 | |||
| BC010070.2 | - | 10 | 1737 | |||
| BC021277.2 | - | 9 | 1631 | 4857987 | ||

TMEM39A
Genomic Map