siRNA Details
| Gene Symbol: CCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002981.2 | NP_002972.1 | 2 | 214 | ||
| GenBank mRNA | AK312183.1 | - | 2 | 212 | ||
| BC105073.1 | - | 2 | 179 | 8144076 | ||
| BC105075.1 | - | 2 | 179 | 8144078 | ||
| DA620119.1 | - | 2 | 212 | |||
| M57502.1 | - | 2 | 192 | |||

CCL1
Genomic Map