siRNA Details
| Gene Symbol: ZNF654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018293.2 | NP_060763.2 | 2 | 2214 | ||
| XM_017006789.1 | XP_016862278.1 | 8 | 4343 | |||
| XM_017006790.1 | XP_016862279.1 | 9 | 3759 | |||
| XM_017006792.1 | XP_016862281.1 | 7 | 3399 | |||
| XM_017006793.1 | XP_016862282.1 | 8 | 3618 | |||
| XM_017006794.1 | XP_016862283.1 | 7 | 3358 | |||
| XM_017006795.1 | XP_016862284.1 | 8 | 3552 | |||
| GenBank mRNA | AF543494.1 | - | 2 | 2216 | ||
| AK001859.1 | - | 2 | 2001 | |||
| AK024795.1 | - | 2 | 878 | |||

ZNF654
Genomic Map