siRNA Details
Gene Symbol: BLOC1S4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018366.2 | NP_060836.1 | 1 | 1320 | ||
GenBank mRNA | AK002092.1 | - | 1 | 1320 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BLOC1S4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018366.2 | NP_060836.1 | 1 | 1320 | ||
GenBank mRNA | AK002092.1 | - | 1 | 1320 |