siRNA Details
| Gene Symbol: C19orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308277.1 | NP_001295206.1 | 8 | 931 | ||
| NM_018381.3 | NP_060851.2 | 8 | 1039 | |||
| XM_011528121.1 | XP_011526423.1 | 10 | 1209 | |||
| XM_017026934.1 | XP_016882423.1 | 8 | 977 | |||
| GenBank mRNA | AK002148.1 | - | 8 | 825 | ||
| AK096142.1 | - | 8 | 1054 | |||
| AK098062.1 | - | 1 | 567 | |||
| AL137694.1 | - | 7 | 863 | |||
| BC035817.2 | - | 8 | 723 | 5554801 | ||

C19orf66
Genomic Map