siRNA Details
| Gene Symbol: EMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018447.3 | NP_060917.1 | 5, 6 | 875 | ||
| XM_005265321.3 | XP_005265378.1 | 5, 6 | 743 | |||
| GenBank mRNA | AF157321.1 | - | 4, 5 | 587 | ||
| AK172843.1 | - | 4, 5 | 583 | |||
| AK222953.1 | - | 4, 5 | 585 | |||
| AK312008.1 | - | 4, 5 | 582 | |||
| BC022807.1 | - | 4, 5 | 570 | 4897577 | ||

EMC3
Genomic Map