siRNA Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018471.2 | NP_060941.2 | 10 | 1554 | ||
| XM_017004502.1 | XP_016859991.1 | 10 | 1424 | |||
| GenBank mRNA | AF109366.1 | - | 10 | 1414 | ||
| AF147724.1 | - | 1 | 169 | |||
| AF161421.1 | - | 10 | 1425 | |||
| AF220184.1 | - | 10 | 1552 | |||
| AF462448.1 | - | 1 | 1128 | |||
| AK001901.1 | - | 10 | 1459 | |||
| AL049948.1 | - | 1 | 450 | |||
| BC021102.1 | - | 10 | 1366 | 5020890 | ||

ZC3H15
Genomic Map