siRNA Details
| Gene Symbol: MIEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019008.5 | NP_061881.2 | 6 | 2709 | ||
| NR_130789.1 | 6 | 2623 | ||||
| XM_011530236.1 | XP_011528538.1 | 8 | 2777 | |||
| XM_011530237.1 | XP_011528539.1 | 7 | 2897 | |||
| XM_011530238.1 | XP_011528540.1 | 7 | 2981 | |||
| XM_017028838.1 | XP_016884327.1 | 7 | 2875 | |||
| GenBank mRNA | AK000239.1 | - | 3 | 1880 | ||
| AK026539.1 | - | 5 | 1296 | |||
| AL365515.1 | - | 6 | 2586 | |||
| AL834205.1 | - | 6 | 2551 | |||
| BC002587.2 | - | 2 | 1851 | |||
| BC008327.1 | - | 6 | 2563 | 3502711 | ||

MIEF1
Genomic Map