siRNA Details
| Gene Symbol: CXCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002994.4 | NP_002985.1 | 3 | 420 | ||
| GenBank mRNA | AK312128.1 | - | 3 | 368 | ||
| BC008376.1 | - | 3 | 371 | 3826939 | ||
| BT006807.1 | - | 3 | 272 | |||
| BU169770.1 | - | 3 | 391 | |||
| CR457428.1 | - | 3 | 272 | |||
| X78686.1 | - | 3 | 378 | |||

CXCL5
Genomic Map