siRNA Details
| Gene Symbol: SHROOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020859.3 | NP_065910.3 | 5 | 1860 | ||
| XM_005263163.4 | XP_005263220.1 | 4 | 6314 | |||
| XM_005263164.4 | XP_005263221.1 | 4 | 872 | |||
| XM_005263165.4 | XP_005263222.1 | 4 | 2298 | |||
| XM_006714282.3 | XP_006714345.1 | 5 | 1015 | |||
| XM_011532157.2 | XP_011530459.2 | 4 | 1702 | |||
| XM_011532158.2 | XP_011530460.1 | 6 | 1661 | |||
| XM_011532159.1 | XP_011530461.1 | 5 | 1850 | |||
| XM_011532160.1 | XP_011530462.1 | 3 | 862 | |||
| XM_011532161.2 | XP_011530463.1 | 4 | 1040 | |||
| XM_017008488.1 | XP_016863977.1 | 5 | 1018 | |||
| XM_017008489.1 | XP_016863978.1 | 5 | 919 | |||
| GenBank mRNA | AB055660.1 | - | 5 | 1861 | ||
| AK128677.1 | - | 1 | 362 | |||
| AK304103.1 | - | 4 | 656 | |||

SHROOM3
Genomic Map