siRNA Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292017.1 | NP_001278946.1 | 3 | 602 | ||
| NM_021204.4 | NP_067027.1 | 3 | 631 | |||
| GenBank mRNA | AF087881.1 | - | 3 | 501 | ||
| AF113125.1 | - | 3 | 502 | |||
| AF177286.1 | - | 1 | 68 | |||
| AK022656.1 | - | 3 | 497 | |||
| BC065815.1 | - | 3 | 469 | 6526329 | ||
| CN335976.1 | - | 3 | 443 | |||
| CR457141.1 | - | 3 | 281 | |||
| DA347866.1 | - | 3 | 538 | |||

ENOPH1
Genomic Map