siRNA Details
| Gene Symbol: AFAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134647.1 | NP_001128119.1 | 18 | 6472 | ||
| NM_198595.2 | NP_940997.1 | 17 | 6220 | |||
| XM_006713909.3 | XP_006713972.1 | 17 | 6103 | |||
| XM_011513544.2 | XP_011511846.1 | 18 | 6525 | |||
| XM_017008535.1 | XP_016864024.1 | 17 | 6003 | |||
| GenBank mRNA | AF188700.1 | - | 16 | 5945 | ||
| D25248.1 | - | 2 | 3797 | |||

AFAP1
Genomic Map