siRNA Details
| Gene Symbol: FASTKD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021826.4 | NP_068598.1 | 2 | 686 | ||
| GenBank mRNA | AB058695.1 | - | 1 | 3052 | ||
| AK023211.1 | - | 2 | 665 | |||
| AK301748.1 | - | 2 | 638 | |||
| BC007413.2 | - | 2 | 686 | 3010170 | ||
| BC011856.1 | - | 2 | 650 | 4298806 | ||

FASTKD5
Genomic Map