siRNA Details
| Gene Symbol: THADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083953.1 | NP_001077422.1 | 11 | 1717 | ||
| NM_001271643.1 | NP_001258572.1 | 11 | 1717 | |||
| NM_001271644.1 | NP_001258573.1 | 11 | 1505 | |||
| NM_022065.4 | NP_071348.3 | 11 | 1505 | |||
| NR_073394.1 | 11 | 1497 | ||||
| XM_006712061.3 | XP_006712124.1 | 12 | 1552 | |||
| XM_006712062.1 | XP_006712125.1 | 11 | 1505 | |||
| XM_006712063.1 | XP_006712126.1 | 11 | 1385 | |||
| XM_006712064.2 | XP_006712127.1 | 11 | 1505 | |||
| XM_006712065.1 | XP_006712128.1 | 11 | 1505 | |||
| XM_006712066.1 | XP_006712129.1 | 11 | 1505 | |||
| XM_006712067.1 | XP_006712130.1 | 11 | 1505 | |||
| XM_006712068.3 | XP_006712131.1 | 11 | 1505 | |||
| XM_011533015.2 | XP_011531317.1 | 11 | 1505 | |||
| XM_017004674.1 | XP_016860163.1 | 11 | 1385 | |||
| XM_017004675.1 | XP_016860164.1 | 11 | 1468 | |||
| GenBank mRNA | AB051554.2 | - | 11 | 1717 | ||
| AK126004.1 | - | 11 | 1420 | |||
| AK126824.1 | - | 1 | 114 | |||
| AK290023.1 | - | 11 | 1505 | |||
| AL832141.1 | - | 11 | 1490 | |||
| AY149629.1 | - | 11 | 1497 | |||
| AY149633.1 | - | 11 | 1489 | |||
| BC037990.2 | - | 11 | 1482 | |||
| BX641038.1 | - | 12 | 1514 | |||
| BX648155.1 | - | 11 | 1501 | |||

THADA
Genomic Map