siRNA Details
| Gene Symbol: ZNF667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321355.1 | NP_001308284.1 | 6 | 2950 | ||
| NM_001321356.1 | NP_001308285.1 | 7 | 3451 | |||
| NM_022103.3 | NP_071386.3 | 5 | 2849 | |||
| XM_011527208.1 | XP_011525510.1 | 6 | 5095 | |||
| XM_011527209.1 | XP_011525511.1 | 5 | 4609 | |||
| GenBank mRNA | BC136388.1 | - | 4 | 2685 | 9020375 | |
| BC150593.1 | - | 4 | 2684 | 9056963 | ||
| BX647401.1 | - | 5 | 2822 | |||
| CR936605.1 | - | 7 | 2762 | |||

ZNF667
Genomic Map