siRNA Details
| Gene Symbol: RTP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022147.2 | NP_071430.2 | 2 | 509 | ||
| GenBank mRNA | AJ251832.1 | - | 2 | 501 | ||
| AY562238.1 | - | 2 | 447 | |||
| BC013161.1 | - | 2 | 509 | 3919961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTP4| Gene Symbol: RTP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022147.2 | NP_071430.2 | 2 | 509 | ||
| GenBank mRNA | AJ251832.1 | - | 2 | 501 | ||
| AY562238.1 | - | 2 | 447 | |||
| BC013161.1 | - | 2 | 509 | 3919961 | ||