siRNA Details
| Gene Symbol: NCAPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022346.4 | NP_071741.2 | 15 | 2456 | ||
| NR_073124.1 | 14 | 2346 | ||||
| XM_011513876.2 | XP_011512178.1 | 14 | 2255 | |||
| XM_017008543.1 | XP_016864032.1 | 15 | 2346 | |||
| XM_017008544.1 | XP_016864033.1 | 14 | 2211 | |||
| GenBank mRNA | AB013299.1 | - | 15 | 2330 | ||
| AF235023.1 | - | 15 | 2259 | |||
| AF331796.1 | - | 15 | 2367 | |||
| AK022512.1 | - | 15 | 2287 | |||
| AK023147.1 | - | 14 | 2037 | |||
| AK027511.1 | - | 6 | 793 | |||
| BC000827.1 | - | 4 | 544 | |||
| BC068467.1 | - | 15 | 2273 | 4821684 | ||
| BC101476.1 | - | 15 | 2244 | 8068982 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019438, AJ237585, AK136254, BC137834, BC144960, and others.

NCAPG
Genomic Map