siRNA Details
Gene Symbol: TMEM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024334.2 | NP_077310.1 | 10, 11 | 1147 | ||
XM_017007176.1 | XP_016862665.1 | 11, 12 | 1269 | |||
GenBank mRNA | AK027466.1 | - | 4, 5 | 377 | ||
AK027757.1 | - | 10, 11 | 1073 | |||
AK027827.1 | - | 5, 6 | 430 | |||
AK027877.1 | - | 10, 11 | 1011 | |||
AK074073.1 | - | 9 | 3578 | |||
AK075010.1 | - | 10, 11 | 998 | |||
AK225589.1 | - | 10, 11 | 1070 | |||
AL136916.1 | - | 10, 11 | 1041 | |||
AY358625.1 | - | 10, 11 | 910 | |||
BC003125.1 | - | 10, 11 | 1042 | 3503636 | ||
BC008054.2 | - | 8, 9 | 637 | |||
BC011719.2 | - | 10, 11 | 1042 | 4079766 | ||
KU178769.1 | - | 9, 10 | 893 | |||
KU178770.1 | - | 8, 9 | 1467 |
