siRNA Details
| Gene Symbol: CEP97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303401.1 | NP_001290330.1 | 3 | 381 | ||
| NM_024548.3 | NP_078824.2 | 3 | 381 | |||
| XM_006713743.3 | XP_006713806.1 | 3 | 874 | |||
| XM_011513127.1 | XP_011511429.1 | 3 | 316 | |||
| XM_017007178.1 | XP_016862667.1 | 3 | 876 | |||
| GenBank mRNA | AK093100.1 | - | 1 | 24 | ||
| AK294784.1 | - | 3 | 319 | |||
| AK309806.1 | - | 3 | 382 | |||
| AL833269.1 | - | 3 | 331 | |||
| BC041085.1 | - | 3 | 324 | 5528397 | ||
| DA520309.1 | - | 3 | 383 | |||

CEP97
Genomic Map