siRNA Details
| Gene Symbol: CEP97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303401.1 | NP_001290330.1 | 11 | 3043 | ||
| NM_024548.3 | NP_078824.2 | 11 | 3220 | |||
| XM_006713743.3 | XP_006713806.1 | 10 | 3611 | |||
| XM_011513127.1 | XP_011511429.1 | 11 | 3177 | |||
| XM_017007178.1 | XP_016862667.1 | 10 | 3436 | |||
| GenBank mRNA | AK026700.1 | - | 1 | 995 | ||
| AK225573.1 | - | 1 | 995 | |||
| AL833269.1 | - | 11 | 3170 | |||
| BC041085.1 | - | 11 | 3163 | 5528397 | ||

CEP97
Genomic Map