siRNA Details
| Gene Symbol: FASTKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281476.2 | NP_001268405.1 | 2 | 745 | ||
| NM_001322046.1 | NP_001308975.1 | 2 | 1418 | |||
| NM_001322048.1 | NP_001308977.1 | 2 | 742 | |||
| NM_001322049.1 | NP_001308978.1 | 2 | 745 | |||
| NM_024622.5 | NP_078898.3 | 2 | 745 | |||
| NR_104020.2 | 2 | 745 | ||||
| XM_006712750.3 | XP_006712813.1 | 2 | 740 | |||
| XM_017004911.1 | XP_016860400.1 | 2 | 1409 | |||
| GenBank mRNA | AK055892.1 | - | 2 | 656 | ||
| AK074302.1 | - | 2 | 636 | |||
| AK130797.1 | - | 2 | 668 | |||
| AL832058.1 | - | 2 | 628 | |||
| BC017950.1 | - | 2 | 636 | |||
| BC032687.2 | - | 1 | 74 | 5581443 | ||

FASTKD1
Genomic Map