siRNA Details
| Gene Symbol: CLIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287527.1 | NP_001274456.1 | 7 | 946 | ||
| NM_001287528.1 | NP_001274457.1 | 7 | 982 | |||
| NM_024692.5 | NP_078968.3 | 7 | 950 | |||
| NR_109844.1 | 6 | 888 | ||||
| XM_005264562.2 | XP_005264619.1 | 7 | 977 | |||
| XM_005264563.1 | XP_005264620.1 | 7 | 982 | |||
| XM_006712104.2 | XP_006712167.1 | 7 | 1022 | |||
| XM_006712105.3 | XP_006712168.1 | 7 | 864 | |||
| XM_011533109.1 | XP_011531411.1 | 7 | 730 | |||
| XM_011533110.1 | XP_011531412.1 | 6 | 783 | |||
| XM_017004958.1 | XP_016860447.1 | 7 | 809 | |||
| XM_017004959.1 | XP_016860448.1 | 7 | 864 | |||
| GenBank mRNA | AB209042.1 | - | 7 | 2561 | ||
| AF433661.1 | - | 7 | 958 | |||
| AK024722.1 | - | 4 | 393 | |||
| AK057267.1 | - | 6 | 889 | |||
| AK225074.1 | - | 4 | 393 | |||
| AK291595.1 | - | 7 | 1027 | |||
| AK302325.1 | - | 7 | 932 | |||
| AK304384.1 | - | 6 | 776 | |||
| AL834443.1 | - | 7 | 924 | |||
| BC013862.1 | - | 7 | 930 | |||
| BC015310.1 | - | 5 | 556 | |||
| BC126167.1 | - | 6 | 720 | 8991883 | ||
| BC136331.1 | - | 6 | 720 | 9020318 | ||

CLIP4
Genomic Map