siRNA Details
| Gene Symbol: NHEJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024782.2 | NP_079058.1 | 8 | 1608 | ||
| GenBank mRNA | AJ972687.1 | - | 8 | 1552 | ||
| AK022672.1 | - | 8 | 1544 | |||
| BC008210.2 | - | 4 | 924 | |||
| BC012732.2 | - | 8 | 1529 | 4026495 | ||

NHEJ1
Genomic Map