siRNA Details
| Gene Symbol: PODNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146254.1 | NP_001139726.1 | 8 | 1770 | ||
| NM_001146255.1 | NP_001139727.1 | 6 | 1503 | |||
| NM_024825.3 | NP_079101.3 | 8 | 1794 | |||
| XM_017027313.1 | XP_016882802.1 | 7 | 1651 | |||
| GenBank mRNA | AK027100.1 | - | 6 | 1494 | ||
| AK290731.1 | - | 8 | 1727 | |||
| AK296101.1 | - | 6 | 1490 | |||
| AK298489.1 | - | 8 | 1770 | |||
| BC057786.1 | - | 8 | 1794 | 30347370 | ||

PODNL1
Genomic Map