siRNA Details
| Gene Symbol: ZNF671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321375.1 | NP_001308304.1 | 2 | 263 | ||
| NM_001321376.1 | NP_001308305.1 | 4 | 428 | |||
| NM_024833.2 | NP_079109.2 | 3 | 393 | |||
| XM_017027314.1 | XP_016882803.1 | 3 | 886 | |||
| GenBank mRNA | AK027159.1 | - | 3 | 381 | ||
| AK091421.1 | - | 4 | 428 | |||
| AK225665.1 | - | 3 | 390 | |||
| AK293680.1 | - | 2 | 261 | |||
| AK310074.1 | - | 2 | 254 | |||
| BC025728.1 | - | 3 | 340 | 5202908 | ||
| DB104548.1 | - | 3 | 393 | |||

ZNF671
Genomic Map