siRNA Details
Gene Symbol: MAP6D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024871.2 | NP_079147.1 | 3 | 1059 | ||
GenBank mRNA | AK022810.1 | - | 3 | 1031 | ||
BC006434.1 | - | 3 | 1024 | 3937613 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MAP6D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024871.2 | NP_079147.1 | 3 | 1059 | ||
GenBank mRNA | AK022810.1 | - | 3 | 1031 | ||
BC006434.1 | - | 3 | 1024 | 3937613 |