siRNA Details
| Gene Symbol: NUP210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024923.3 | NP_079199.2 | 21 | 3011 | ||
| XM_017006018.1 | XP_016861507.1 | 3 | 844 | |||
| XM_017006019.1 | XP_016861508.1 | 3 | 811 | |||
| XM_017006020.1 | XP_016861509.1 | 3 | 2883 | |||
| GenBank mRNA | AB020713.1 | - | 1 | 27 | ||
| AK074101.1 | - | 6 | 633 | |||

NUP210
Genomic Map