siRNA Details
| Gene Symbol: NUP210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024923.3 | NP_079199.2 | 40 | 5995 | ||
| XM_017006018.1 | XP_016861507.1 | 22 | 3828 | |||
| XM_017006019.1 | XP_016861508.1 | 22 | 3795 | |||
| XM_017006020.1 | XP_016861509.1 | 22 | 5867 | |||
| GenBank mRNA | AB020713.1 | - | 20 | 3011 | ||
| AK074101.1 | - | 25 | 3617 | |||
| AK075545.1 | - | 10 | 1690 | |||
| AL117527.1 | - | 9 | 1542 | |||

NUP210
Genomic Map