siRNA Details
| Gene Symbol: TMEM156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303228.1 | NP_001290157.1 | 3 | 533 | ||
| NM_024943.2 | NP_079219.1 | 3 | 533 | |||
| XM_005262670.3 | XP_005262727.1 | 4 | 583 | |||
| XM_011513749.2 | XP_011512051.1 | 4 | 580 | |||
| XM_011513750.2 | XP_011512052.1 | 4 | 581 | |||
| XM_011513751.2 | XP_011512053.1 | 4 | 584 | |||
| XM_011513752.2 | XP_011512054.1 | 4 | 584 | |||
| XM_011513753.2 | XP_011512055.1 | 4 | 498 | |||
| XM_011513754.2 | XP_011512056.1 | 3 | 796 | |||
| XM_017008629.1 | XP_016864118.1 | 4 | 579 | |||
| GenBank mRNA | AK026888.1 | - | 3 | 533 | ||
| BC030803.1 | - | 3 | 474 | 4806992 | ||
| BC060512.1 | - | 3 | 499 | |||

TMEM156
Genomic Map