siRNA Details
| Gene Symbol: CSRNP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172173.1 | NP_001165644.1 | 6 | 830 | ||
| NM_024969.3 | NP_079245.2 | 4 | 729 | |||
| XM_005246865.4 | XP_005246922.1 | 4 | 552 | |||
| GenBank mRNA | AB063300.1 | - | 4 | 790 | ||
| AK056655.1 | - | 4 | 726 | |||
| AK126129.1 | - | 6 | 828 | |||

CSRNP3
Genomic Map