siRNA Details
| Gene Symbol: THOC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285387.2 | NP_001272316.1 | 7 | 632 | ||
| NM_001285404.1 | NP_001272333.1 | 8 | 881 | |||
| NM_025075.3 | NP_079351.2 | 8 | 750 | |||
| XM_006713339.3 | XP_006713402.1 | 8 | 699 | |||
| GenBank mRNA | AF104914.1 | - | 7 | 598 | ||
| AK027098.1 | - | 8 | 660 | |||
| AK225110.1 | - | 8 | 638 | |||
| BC020599.2 | - | 7 | 598 | |||
| BC065012.1 | - | 8 | 620 | 6167514 | ||

THOC7
Genomic Map