siRNA Details
| Gene Symbol: COQ10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320818.1 | NP_001307747.1 | 4 | 1574 | ||
| NM_001320819.1 | NP_001307748.1 | 5 | 1768 | |||
| NM_001320820.1 | NP_001307749.1 | 5 | 1965 | |||
| NM_025147.4 | NP_079423.1 | 5 | 1724 | |||
| GenBank mRNA | AK023510.1 | - | 5 | 1593 | ||
| BC022453.1 | - | 5 | 1641 | 4796819 | ||

COQ10B
Genomic Map