siRNA Details
| Gene Symbol: COQ10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320818.1 | NP_001307747.1 | 2, 3 | 429 | ||
| NM_001320819.1 | NP_001307748.1 | 3, 4 | 623 | |||
| NM_001320820.1 | NP_001307749.1 | 3, 4 | 820 | |||
| NM_025147.4 | NP_079423.1 | 3, 4 | 579 | |||
| GenBank mRNA | AK023510.1 | - | 3, 4 | 448 | ||
| AK294093.1 | - | 3, 4 | 820 | |||
| AK308977.1 | - | 3, 4 | 417 | |||
| BC022453.1 | - | 3, 4 | 496 | 4796819 | ||
| BU195968.1 | - | 2, 3 | 363 | |||
| DC328985.1 | - | 3, 4 | 417 | |||

COQ10B
Genomic Map