siRNA Details
| Gene Symbol: TMEM177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105198.1 | NP_001098668.1 | 2 | 838 | ||
| NM_001105199.1 | NP_001098669.1 | 2 | 446 | |||
| NM_030577.2 | NP_085054.2 | 2 | 497 | |||
| GenBank mRNA | AK096890.1 | - | 2 | 443 | ||
| AK130088.1 | - | 2 | 446 | |||
| AK299078.1 | - | 2 | 443 | |||
| BC004404.1 | - | 2 | 485 | 3637432 | ||
| BG280893.1 | - | 2 | 440 | |||
| BG776153.1 | - | 2 | 468 | |||

TMEM177
Genomic Map