siRNA Details
| Gene Symbol: FAM49A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030797.3 | NP_110424.1 | 12 | 1182 | ||
| XM_005262630.3 | XP_005262687.1 | 11 | 1138 | |||
| XM_006712108.3 | XP_006712171.1 | 12 | 1137 | |||
| GenBank mRNA | AK001942.1 | - | 4 | 238 | ||
| AK055334.1 | - | 12 | 1150 | |||
| AK091280.1 | - | 7 | 596 | |||
| AK315065.1 | - | 11 | 1077 | |||
| AL136704.1 | - | 12 | 1147 | |||
| BC038971.1 | - | 12 | 1133 | 5200807 | ||
| CR533504.1 | - | 10 | 929 | |||

FAM49A
Genomic Map