siRNA Details
| Gene Symbol: FGFBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031950.3 | NP_114156.1 | 2 | 1138 | ||
| GenBank mRNA | AB021123.1 | - | 2 | 1109 | ||
| AY358978.1 | - | 2 | 1138 | |||
| BC025720.1 | - | 2 | 1107 | 5228917 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFBP2| Gene Symbol: FGFBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031950.3 | NP_114156.1 | 2 | 1138 | ||
| GenBank mRNA | AB021123.1 | - | 2 | 1109 | ||
| AY358978.1 | - | 2 | 1138 | |||
| BC025720.1 | - | 2 | 1107 | 5228917 | ||