siRNA Details
| Gene Symbol: WDR54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320823.1 | NP_001307752.1 | 8 | 795 | ||
| NM_001320824.1 | NP_001307753.1 | 8 | 850 | |||
| NM_001320825.1 | NP_001307754.1 | 7 | 594 | |||
| NM_032118.3 | NP_115494.1 | 8 | 817 | |||
| XM_017005064.1 | XP_016860553.1 | 7 | 596 | |||
| GenBank mRNA | AK023015.1 | - | 8 | 810 | ||
| AK311621.1 | - | 3 | 396 | |||
| BC051848.1 | - | 8 | 782 | 6150670 | ||
| CD249537.1 | - | 7 | 610 | |||

WDR54
Genomic Map