siRNA Details
| Gene Symbol: WDR54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320823.1 | NP_001307752.1 | 2 | 176 | ||
| NM_001320824.1 | NP_001307753.1 | 2 | 231 | |||
| NM_032118.3 | NP_115494.1 | 2 | 198 | |||
| GenBank mRNA | AK023015.1 | - | 2 | 191 | ||
| AK222696.1 | - | 2 | 191 | |||
| BC051848.1 | - | 2 | 163 | 6150670 | ||
| BG024606.1 | - | 2 | 197 | |||
| BI855898.1 | - | 2 | 139 | |||
| HY088750.1 | - | 2 | 213 | |||

WDR54
Genomic Map