siRNA Details
| Gene Symbol: SLC16A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166496.1 | NP_001159968.1 | 2, 3 | 1057 | ||
| NM_003051.3 | NP_003042.3 | 2, 3 | 588 | |||
| XM_011542026.2 | XP_011540328.1 | 2, 3 | 321 | |||
| XM_011542027.2 | XP_011540329.1 | 2, 3 | 331 | |||
| GenBank mRNA | AK094064.1 | - | 2, 3 | 1057 | ||
| AK312500.1 | - | 2, 3 | 431 | |||
| AL162079.1 | - | 2, 3 | 418 | |||
| BC026317.1 | - | 2, 3 | 431 | 4811581 | ||
| BC045664.1 | - | 3, 4 | 1142 | 5298234 | ||
| L31801.1 | - | 1, 2 | 237 | |||

SLC16A1
Genomic Map