siRNA Details
| Gene Symbol: WDR75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303096.1 | NP_001290025.1 | 6, 7 | 597 | ||
| NM_032168.2 | NP_115544.1 | 5, 6 | 563 | |||
| GenBank mRNA | AK022581.1 | - | 5, 6 | 510 | ||
| AK022607.1 | - | 5, 6 | 522 | |||
| AK023276.1 | - | 2, 3 | 202 | |||
| AK091546.1 | - | 6, 7 | 524 | |||
| AK290445.1 | - | 5, 6 | 514 | |||
| AL122097.1 | - | 1, 2 | 130 | |||
| BC040567.1 | - | 5, 6 | 563 | 5267120 | ||
| BI460450.1 | - | 5, 6 | 568 | |||
| CR749440.1 | - | 5, 6 | 2172 | |||

WDR75
Genomic Map